Iodine 0.025 M (0.05 N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Warning

85805.260EA 0 INR
85805.260
Iodine 0.025 M (0.05 N) aqueous solution, AVS TITRINORM Volumetric
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
Merck Index: 13,05036

Order Now

Specification Test Results

Titer (20°C; real value 0.5 % accuracy) 0,02488 --> 0,02512 mol/l

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR