VWR®, Melting point standards
Supplier: VWR Chemicals
85107.001EA 9011.55 INR
85107.001 VWRC85105.005_U 85106.001 85109.001 85110.001 85111.003 85112.003 85113.003 85101.001 85103.001 85104.001
VWR®, Melting point standards
Standards Melting Point Standards
These standards can be used with any melting point apparatus and are prepared using pure raw materials. Melting points are determined using a high accuracy Differential Scanning Calorimeter (DSC) system that is calibrated to the ITS 90 International Temperature Scale.
  • Ready to use
  • Traceable
  • Uncertainty of measurement up to ±0,3 °C
The melting points of these standards range from benzophenone (47 to 49 °C) to anthraquinone (283 to 286 °C).