2-Propanol 99,9, HiPerSolv CHROMANORM® LC-MS

Supplier: VWR Chemicals
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

Filter 0,2 um_Nitrogen_filled

84881.320
84881.320EA 28861.55 INR
84881.320 84881.290
2-Propanol 99,9, HiPerSolv CHROMANORM® LC-MS
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) ≥ 99,9 %
Acidity ≤ 0,0002 meq/g
Alkalinity ≤ 0,0002 meq/g
Evaporation residue ≤ 1 ppm
Water ≤ 50 ppm
Ag (Silver) ≤ 10 ppb
Al (Aluminium) ≤ 25 ppb
Ca (Calcium) ≤ 25 ppb
Cu (Copper) ≤ 10 ppb
Fe (Iron) ≤ 10 ppb
K (Potassium) ≤ 10 ppb
Mg (Magnesium) ≤ 10 ppb
Mn (Manganese) ≤ 10 ppb
Na (Sodium) ≤ 50 ppb
Ni (Nickel) ≤ 10 ppb
Pb (Lead) ≤ 10 ppb
Zn (Zinc) ≤ 10 ppb
Transmittance (205 nm) ≥ 10 %
Transmittance (210 nm) ≥ 50 %
Transmittance (225 nm) ≥ 80 %
Transmittance (240 nm) ≥ 95 %
Transmittance (255 nm) ≥ 99 %
Suitable for LC-MS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR