Labelling tapes, industrial grade vinyl tape
Supplier: MICRONOVA
47B-12RE
817-0224EA 0 INR
817-0224
Labelling tapes, industrial grade vinyl tape
Tapes
Processed in a controlled environment, tapes are wound on a plastic core and single bagged. Ideal for wheel taping and colour coding applications such as spray bottle identification.
  • Forms strong bond without leaving adhesive residue
  • Can be written on with cleanroom pen
  • Different colours available
Length: 32,9 m