Acetone, BAKER ANALYZED® HPLC HPLC, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

8142.2500EA 0 INR
8142.2500
Acetone, BAKER ANALYZED® HPLC HPLC, J.T.Baker®
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) (corrected for water) min. 99.7 %
Aldehyde (as HCHO) max. 0.002 %
Color (APHA) max. 10
Isopropyl Alcohol max. 0.05 %
Methanol (CH3OH) max. 0.05 %
Residue after Evaporation max. 2 ppm
Solubility in Water passes test
Substances Reducing KMnO4 passes test
Titrable Acid (meq/g) max. 0.0003
Titrable Base (meq/g) max. 0.0006
Water (H2O) max. 0.2 %
Ultraviolet Absorbance (1.00-cm path vs water):
at 330 nm max. 1.00
at 340 nm max. 0.10
at 350 nm max. 0.01
at 400 nm max. 0.01
UV Cut-off, nm max. 330


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR