Sodium hydrogen carbonate 99,7-100,3 Acidimetry, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Sodium bicarbonate

7412-09EA 261151.07 INR
7412-09 7412-20 MACR7412-12 MACR7412-06
Sodium hydrogen carbonate 99,7-100,3 Acidimetry, AR® ACS, Macron Fine Chemicals™
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.7 - 100.3 %
ACS - Assay (NaHCO₃) (by acidimetry) 99.7 - 100.3 %
ACS - Ammonium (NH₄) ≤5 ppm
ACS - Calcium (Ca) ≤0.02 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.015 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Potassium (K) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR