VWR®, Cell Slides in Sulture Dishes
Supplier: VWR Collection
734-2973EA 20822.76 INR
734-2973 734-2974 734-2975 734-2976
VWR®, Cell Slides in Sulture Dishes
Culture Dishes Cell Culture Dishes
VWR® cell slides are made from medical grade, high transparency PET, which has high strength and is unbreakable. Only the surface of the cell slide has a hydrophilic treatment so adherent cells grow easily, whilst the surrounding culture dish surface is hydrophobic, making it difficult for adherent cells to grow.
  • Cell slide transparency and light transmittance are good so cells can be clearly observed under a light or fluorescence microscope
  • The use of multiple cell slides in one dish makes it possible to develop multi-factor, multi-index, multi-level in
More Product Information