VWR®, Single-Use Tank Liners, Medical Grade
Supplier: VWR Collection
VWRI734-2950_SEA 0 INR
VWRI734-2950_S VWRI734-2952_S 734-2943 734-2944 734-2945 734-2946 734-2947 734-2948 734-2949 734-2950 734-2951 734-2952 734-2953 734-2954
VWR®, Single-Use Tank Liners, Medical Grade
Bags Media Bags
These tank liners are single-use bags with an open top, designed to fit in cylindrical vessels.
  • No cleaning after media preparation
  • Medical grade, multi-layer film
  • For bioprocess applications
  • Delivered gamma irradiated and ready-to-use
These bags are used as a liner into a vessel to avoid cleaning of the vessel after media and buffer preparation. After installing the Tank Liner into a vessel and filling it with liquid, powder can be added to the vessel and the mixing process can start by using an overhead agitator. When mixing is finished, it’sMore Product Information