UV transilluminators, high performance
Supplier: UVP ULTRA VIOLET PRODUCTS
95-0421-02 95-0422-02 95-0423-02 95-0424-02 95-0425-02 95-0427-02 95-0428-02 95-0429-02 95-0431-02 95-0432-02
732-4375EA 0 INR
732-4375 732-4376 732-4377 732-4378 732-4379 732-4381 732-4382 732-4383 732-4385 732-4386
UV transilluminators, high performance
Transilluminators UV Transilluminators
These High Performance UV transilluminators, with four 25 W tubes, provide 100 W of high intensity and uniform UV illumination for high fluorescence and sensitivity when viewing stained gels. Hi/Med/Lo intensity settings (on selected models) are for UV illumination, fluorescence viewing and photography applications. The 'Lo' intensity setting reduces photonicking or photobleaching of gel samples during prep work, allowing for extended gel preparation time. The 'Hi' and 'Med' intensity settings are used in analytical documentation work.
  • Stainless steel frame enables easy cleaning and
More Product Information