VWR®, Thermal Cyclers
Supplier: VWR Collection
732-2548EA 1063374.48 INR
732-2548 732-2549 732-2550 732-2551 732-2552 732-2554 732-2555 732-2914 732-2915
VWR®, Thermal Cyclers
PCR Cyclers
The VWR® thermal cycler family combines high quality engineering with a comprehensive range of block formats. The UNO cycler is designed around a powerful, yet easy to use, software interface, and uses the same platform for both 96-well and 384-well formats. The Doppio, with two independent high speed 48-well blocks in one system, offers the optimum solution for maximum flexibility within a minimum footprint. Because of the special design of the heated lid, the height adjusts automatically to the different tube sizes.
  • Outstanding performance: Thermal plate with impressive thermal
More Product Information