VWR®, UV Transilluminators for VWR® Imager System
Supplier: VWR Collection
730-1461EA 122768.52 INR
730-1461 730-1462 730-1463 730-1480 730-1481 730-1472 730-1473 730-1474 730-1475 730-1476 730-1477 730-1478 730-1479 730-1483 730-1482
VWR®, UV Transilluminators for VWR® Imager System
Transilluminators UV Transilluminators
All VWR® transilluminators slide out for easy access to provide very even illumination. Those for use with the VWR® Imager system also have variable intensity settings.
  • Choice of filter sizes
  • Safety cut-off (when in darkroom)
  • Choice of different/multiple wavelengths available
  • Available in Benchtop version with protective lid
Order Now
SPECIFICATIONS
Environmentally Preferable