VWR® Q Path® MacroStar VII, Cassettes
Supplier: VWR Collection
720-2207EA 0 INR
720-2207 720-2208 720-2209 720-2210 720-2211 720-2213 720-2214 720-2206 720-2212
VWR® Q Path® MacroStar VII, Cassettes
Embedding Cassettes Biopsy Processing Cassettes
Suitable for holding and identifying large tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Supplied in dispensers containing 500 cassettes. Pack of 2 dispensers (1000 cassettes). Double the height of standard cassettes.
  • Large cassette: 32×26×12 mm
  • Welded lid
  • 45° writing angle
Order Now
SPECIFICATIONS
Environmentally Preferable