VWR® Q Path® NovaBrush® Cytological Endo- and Exocervical Brushes
Supplier: VWR Collection
720-2066EA 0 INR
720-2066 720-2069 720-2067 720-2068 720-2070 720-2071
VWR® Q Path® NovaBrush® Cytological Endo- and Exocervical Brushes
Brushes
Q Path® NovaBrush® are disposable brushes designed for endo- and exocervical cytological sampling. The shape of Q Path® NovaBrush® 1's head makes it ideal for endocervical sampling without risk of trauma to the patient. With an undetachable handle (pink), the single-use Q Path® NovaBrush® 1 is ideal for conventional Pap smear testing and thin layer cytology. Q Path® NovaBrushes 2 and 3 are suitable for both endo- and exocervical sampling. They have ergonomically-designed round pink (undetachable), or green (detachable) handles. Q Path® NovaBrush 2 and 3 are recommended for liquid-basedMore Product Information