Tissue processing/embedding cassettes, Histosette® II, M492
Supplier: SIMPORT SCIENTIFIC
M492-12
720-0908EA 0 INR
720-0908
Tissue processing/embedding cassettes, Histosette® II, M492
Embedding Cassettes Tissue Processing Cassettes
Acetal polymer.
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
  • Integral lid
  • Anterior writing area at 45° angle to enable use with leading cassette labelling instruments