VWR® SAS Super DUO 360 Air Samplers
Supplier: VWR Collection
710-0867EA 344285.94 INR
710-0867 710-0866
VWR® SAS Super DUO 360 Air Samplers
Samplers Air Samplers
When environmental contamination is very low, and the number of samples to be performed is high, operational duration can be an important factor in the choice of SAS instrument. Two sampling heads that operate simultaneously to sample 1000 litres of air, in less than 3 minutes, is the solution.
  • Single body with two aspirating heads
  • All manuals are available on a USB key
  • Instruction manual in six different languages
The simultaneous sampling on two heads also allows you to obtain statistically representative results when using the same type of media inMore Product Information