Medium swabs, CleanFoam®
Supplier: ITW-CONTAMINATION TEXWIPE
TX712A
710-0551EA 39783.63 INR
710-0551
Medium swabs, CleanFoam®
Swabs Swabs for Cleaning
Thermally-bonded swab for applications requiring high absorbency, low particle generation, and uniform dispersing of solvents.
  • Inherent softness and low abrasion protect delicate components
Packaging: Cleanroom processed bag-within-a-bag.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904