Swabs with large circular foam head, CleanFoam®
Supplier: ITW-CONTAMINATION TEXWIPE
TX708A
710-0548EA 36728.38 INR
710-0548
Swabs with large circular foam head, CleanFoam®
Swabs Swabs for Cleaning
Thermally-bonded swabs with 100 pore per inch porosity.
  • Head will not crack or chip
  • Inherent softness and low abrasion protect delicate components
  • Large circular head on a long and easy grip handle
Can be used in industrial applications, from general assembly to cleanrooms, requiring high absorbency with low particle generation and uniform dispersing of solvents.
Packaging: Cleanroom processed bag-within-a-bag.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904