Nitric acid (1:1) v/v 38% v/v

Supplier: Hach
Danger

Synonyms: Nitric acid solution

254049
705-1124EA 0 INR
705-1124
Nitric acid (1:1) v/v 38% v/v
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: ∼86 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.2…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR