Luminescent/optical dissolved oxygen (LDO) electrodes, IntelliCAL™
Supplier: HACH
LDO10105 LDO10101 LDO10103 LDO10110 LDO10115 LDO10130
664-0208EA 0 INR
664-0208 664-0206 664-0207 664-0209 664-0210 664-0211
Luminescent/optical dissolved oxygen (LDO) electrodes, IntelliCAL™
Electrodes Dissolved Oxygen Electrodes
IntelliCAL™ LDO101 is a digital, luminescent dissolved oxygen (LDO) electrode which is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
  • Rugged digital probes can reliably test in the harshest environmental conditions: Stainless steel body, polymer sensor shroud, and reinforced, steel-sheathed cables protect the electrode for measurement anywhere in the field
  • No membranes to clean or replace, no electrolyte solution to replenish, and reduce calibration frequency
  • Ultimate traceability: Stored time and
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable