VWR® CO-2500L, Conductivity/salinity/TDS/°C lab meter, with colour touch screen
Supplier: VWR Collection
663-0372EA 58508.94 INR
663-0372 663-0373
VWR® CO-2500L, Conductivity/salinity/TDS/°C lab meter, with colour touch screen
Electrochemistry Meters Conductivity Meters
User friendly instrument distinguished by its large 127 mm colour graphic touch screen. The meter can be used for accurate measurements of conductivity, salinity, TDS, resistivity and temperature. This CO meter enables simultaneous measurements displaying and storing of two functions (conductivity and temperature).
  • Large five colour graphic touch screen showing both conductivity and temperature value simultaneously
  • Wide measuring range for measurements in ultra-pure water as well as in high conductivity solutions
  • Non linear temperature compensation for
More Product Information