Mineral oil light, white NF, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

6358-19EA 0 INR
6358-19 6358-04 6358-10
Mineral oil light, white NF, Macron Fine Chemicals™
Mineral oil
Boiling Pt: 300…500 °C (1013 hPa)
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 215 °C (closed cup)
MDL Number: MFCD00131611
CAS Number: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07093

Order Now

Specification Test Results

Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 0.818 - 0.880
NF - Specific Gravity at 25°/25°C 0.818 - 0.880
NF - Viscosity at 40 C, mm₂/s 3.0 - 34.4
NF - Readily Carbonizable Substances Passes Test
NF - Solid Paraffin Passes Test
NF - Acidity Passes Test
NF - Limit of Polycyclic Aromatic Hydrocarbons Passes Test
NF - Limit of Sulfur Compounds Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR