VWR®, Cuvettes for Fluorescence Measurements
Supplier: VWR Collection
634-1223EA 0 INR
634-1223 634-1224 634-1225
VWR®, Cuvettes for Fluorescence Measurements
Cuvettes Spectrophotometer Cuvettes
Standard rectangular macro cuvettes made from Spectrosil quartz for accurate readings.
  • Fully heat-fused construction
  • Four polished windows
  • Two cover types available: Flat lid and stopper
Due to the fully heat-fused construction (no intermediate materials or adhesives), and careful annealing to remove any strain, there is no optical or physical distortion of the two optically polished windows. Flat lids and stoppers are made of polytetrafluoroethylene (PTFE). Stoppered cells provide a more suitable seal for volatile liquids.
Certifications: More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable