D-(+)-Galactose kit 99 HPLC_ASSAY_METHOD, multi-compendial Endotoxins_low

Supplier: Avantor

Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal

M.O.C. Category R

6322-X1EA 271881.65 INR
6322-X1
D-(+)-Galactose kit 99 HPLC_ASSAY_METHOD, multi-compendial Endotoxins_low
D(+)-Galactose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 527
Density: 1.72 g/cm³
MDL Number: MFCD00151230
CAS Number: 59-23-4
Merck Index: 13,04356

Order Now

Specification Test Results

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Kit Components Meet Requirements Passes Test
This kit contains one each of Galactose 6322-B1 and 6322-SA.
Solvent Ethanol are likely to be present. Each is tested and the
concentration reported for each batch.
Plant derived.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.
Plant Derived
This product has been filled to +/- 1% of the stated net weight.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR