Staining tiles

Supplier: KARTELL
356
631-9143EA 0 INR
631-9143
Staining tiles
Staining Tiles
This PE staining tile has three round depressions of Ø 21 mm.

  • Suitable for colorimetric tests

Colour: white
W×D: 28×85 mm.

W×D: 28×85 mm

Colour: White
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR