VWR® SuperFrost® BS 7011, Microscope Slides
Supplier: VWR Collection
631-0911EA 566 INR
631-0911 631-0912 631-0913 631-0909 631-0910
VWR® SuperFrost® BS 7011, Microscope Slides
Slides Microscope Slides
These microscope slides come with a light opaque coating in different colours.
  • Resistant to commonly used solvents
  • No scratching or sticking when stacked together
  • Label areas on the top of the slide, for quick identification
Each colour codes different specimens at a glance according to urgency, processing or staining technique.
Certifications: BS 7011
Order Now
SPECIFICATIONS
Environmentally Preferable