Gooseneck light guides
Supplier: SCHOTT
154202
631-0022EA 0 INR
631-0022
Gooseneck light guides
Lights Light Guides
Fibre optic illumination for stereo microscopy. To be used with cold light sources.
In all its microscopy light guides, SCHOTT uses the new environmentally friendly glass optical fibers PURAVIS®. Not just the fibers themselves, but also the entire proprietary manufacturing process are lead free and avoid the use of arsenic and antimony. Due to superior physical and chemical properties, PURAVIS® glass optical fibers provide a high performance, superior longevity, improved transmission in the near UV and outstanding transmission of white light.