VWR® VisiScope® 254, Upright Microscopes
Supplier: VWR Collection
630-1928EA 86879.94 INR
630-1928 630-1929 630-2684 630-2688
VWR® VisiScope® 254, Upright Microscopes
Microscopes
These upright microscopes are ideal for use in universities and routine laboratories.
  • Die-cast frame with high stability
  • N-plan objectives (160 mm or IOS)
  • Fixed Koehler illumination for crisp images
  • LED illumination with adjustable intensity; constant pure-white colour temperature (6300 K)
  • Optical components treated to prevent the formation of moulds and fungi
Certifications: Objectives conform to DIN standards.
Strict controls on materials and production processes optimised through the procedures established by the ISO 9001:2008 quality system.