Counting chambers, Bürker
Supplier: MARIENFELD
0640211
630-1541EA 0 INR
630-1541
Counting chambers, Bürker
Slides Counting Chambers
Glass counting chambers equipped with two sets of grids, engraved into two ground, and polished surfaces on the central ridge. This central ridge is located between two elevated ridges, that are also ground and polished.
Designed for investigations of cell suspensions.
Conform to DIN 12847 and the German Calibration Ordinance
Certifications: DIN 12847 and the German Calibration Ordinance.
Delivery information: Supplied in a transparent box.