VWR® Traceable®, Extreme Accuracy Triple-Display Thermometers
Supplier: VWR Collection
620-1824EA 15681.96 INR
620-1824 620-1884 620-1886 620-1885
VWR® Traceable®, Extreme Accuracy Triple-Display Thermometers
Thermometers Digital Thermometers
These international standard digital thermometers provide exact readings necessary for critical requirements, quality control checks and routine measurements. The stainless steel probe shows the slightest temperature change in liquid, air/gas and semi-solids. It delivers exact time/date when temperatures rise above/fall below set points.
  • Three-line display: Maximum/minimum and current temperatures
  • Displays the exact time and date when the limits occurred
  • Programmable alarm provides two visual (LED) and two audio alerts when temperature rises above or falls
More Product Information