VWR® PHP Pipettes, Special Edition
Supplier: VWR Collection
613-7184EA 16527.78 INR
613-7184 613-7185 613-7186 613-7187 613-7188 613-7189 613-7190 613-7191 613-7192 613-7193 613-7194 613-7195 613-7196 613-7197 613-7198 613-7199 613-7200 613-7201 613-7202 613-7203 613-7204 613-7205 613-7206 613-7207 613-7208 613-7209 613-7210 613-7211 613-7212 613-7213 613-7214 613-7215 613-7216 613-7217 613-7218
VWR® PHP Pipettes, Special Edition
Pipettes Micropipettes
VWR® PHP pipettes are fully autoclavable, high performance pipettes with low plunger force.
  • Superior accuracy and precision (tested in ISO 17025 accredited lab)
  • Magnet assisted piston for low plunger force and lesser chance of RSI
  • PVDF tip cone for durable performance
  • Large and clear 4 digit display; color coding for easy identification
  • Three-year warranty
VWR® PHP Pipettes are Precise High Performance, fully autoclavable pipettes, that are available in Single and Multi Channel range. VWR® PHP offers one of the lowest plunger forces and a durable tipMore Product Information