VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
612-7259EA 0 INR
612-7259 612-7260 612-7262 612-7263 612-7264 612-7265 612-7266 612-7268 612-7270 612-7271 612-7272 612-7273 612-7274 612-7277 612-7278
VWR® Inoculating Loops and Needles, Disposable
Spreaders Inoculating Loops and Needles
Inoculating loops and needles are devices used for inoculation, serial dilution, sterile sampling, transfer, and microbiological sample spreading.
  • PS, sterile
  • Colour coded for easy product identification
  • Hexagonal loop shaft improves grip, assisting orientation and making diluting and streaking easier
  • E-beam sterilised
The resealable pouch allows loops to be easily accessed any number of times, then re-closed to maintain product integrity. The loops and needles are positioned upside down in the pouch in order to reduce any risk of contamination.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable