VWR®, Volumetric Pipettes, ISO
Supplier: VWR Collection
612-6875EA 4149.56 INR
612-6875 612-6876 612-6879 612-6884 612-6886 612-6887 612-6888 612-6889 612-6950 612-6951 612-6952 612-6953 612-6954 612-6955 612-6956 612-6957
VWR®, Volumetric Pipettes, ISO
Pipettes Bulb Pipettes
VWR® borosilicate glass pipettes are made from specially selected accurate bore glass tubing, have precision formed jets, bevel ground ends to resist chipping. The mouth piece of the pipettes is also tooled for easy sucking and cotton plugging.
  • Calibrated at 20 °C
  • Calibrated for delivery (TD, Ex)
  • Generally used for accurate measurement and decanting of liquid
  • Markings are made in permanent amber stain diffused into the surface of the glass for better visibility and durability
Each Pipette is calibrated to deliver (Ex, TD) at +20 °C referenceMore Product Information