VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
16001-186 16001-196 16001-172 16001-192 414004-008 16001-170 414004-032 16001-174 414004-028 16001-182 414004-004 414004-018 414004-017 16001-180 16001-188 414004-023
612-4466EA 0 INR
612-4466 612-4518 612-4468 612-4469 612-4470 612-4471 612-4472 612-4473 612-4520 612-4476 612-4479 612-4481 612-4486 612-4490 612-4491 612-4492 612-4494 612-4495 612-4496 612-4497 612-4499 612-4500 612-4502 612-4543 612-4504 612-4505 612-4506 612-4508 612-4509 612-4511 612-4512 612-4513 612-4514 612-4515 612-4542 612-4533 612-4526 612-4527 612-4530 612-4523 612-4536 612-4538 612-4545 VWRU16001-186 VWRU16001-196 612-3399 612-3608 612-3609 612-3610 612-3619 612-3627 612-5550 VWRU16001-192 VWRU414004-008 VWRU16001-170 VWRU414004-032 VWRU16001-174 612-5559 612-5549 VWRU414004-004 VWRU414004-018 612-5551 VWRU16001-180 612-5557 VWRU414004-023 612-5770 612-5781
VWR® Standard Line Transfer Pipettes
Pipettes Pasteur/Transfer Pipettes
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials.
  • Will not shatter or break
  • Non toxic and inert to biological fluids and most acids
  • Can be sealed, refrigerated and gas sterilised
  • No bulb to insert or remove
  • Uniform drop size
The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
Ordering information: To help you find the correct product andMore Product Information