VWR® IS High Capacity Balances
Supplier: VWR Collection
611-4530EA 71103.9 INR
611-4530 611-4549 611-4550 611-4536 611-4528 611-4537 611-4529 611-4539 611-4557 611-4558 611-4559 611-4540 611-4538 611-4556 611-4547 611-4548 611-4520 611-4546 611-4946 611-4947 611-4948 611-4949 611-4950 611-4951
VWR® IS High Capacity Balances
Balances and Scales
The IS range of high capacity balances are robust, reliable and ensure high performance. Models are equipped with either a backlit LCD with adjustable contrast, a large graphic display or a large 5" colour touch screen, the balances have been designed to deliver top performance at competitive prices.
  • Electromagnetic force restoration weighing system (for 0,01 g resolution)
  • High readability load cell weighing system (for 0,1 g resolution)
  • Solidly built with an ergonomic keypad
  • External calibration
  • Measuring units are selectable and customisable
  • Hook for
More Product Information