VWR®, Weighing Boats, Paper
Supplier: VWR Collection
611-3256EA 0 INR
611-3256 611-3257 611-3258 611-3259 611-3624 611-3625
VWR®, Weighing Boats, Paper
Boats Weighing Boats
VWR® paper weighing boats are a first in eco friendly laboratory consumables.
  • Control contents like a polystyrene weighing boat making them easier to handle than standard, flat weighing paper
  • Made of greaseproof paper that meets FDA 21 CFR 176.170, components of paper in contact with aqueous and fatty foods, with 2I CFR 176.180 and contact with dry foods
  • Compatible with heat applications up to a maximum of 210 °C
They are both Eco-In and Eco-Out.
Eco-In because they are FSC® Certified. The papers used to manufacture our boats are made from responsiblyMore Product Information