VWR® Advanced LPG / LPCG / LPWG, Precision Balances with Graphic Display
Supplier: VWR Collection
611-2306EA 65294.46 INR
611-2306 611-2291 611-2297 611-2310 611-2307 611-2285 611-2287 611-2283 611-2373 611-2372 611-3273 611-3274 611-3275 611-3276 611-3278 611-3284 611-3358 611-3365 611-3366 611-3381 611-3390 611-3393 611-3460 611-3474 611-3832 611-3833 611-4318 611-4329 611-4344 611-4345 611-4346 611-4347 611-4348 611-5048
VWR® Advanced LPG / LPCG / LPWG, Precision Balances with Graphic Display
Balances and Scales
Precision balances that are ideal for use in any laboratory, these models offer GLP/ISO records of weight values with date, time and serial number. A series that ranges from simple weighing to operation with automated internal calibration. All Advanced models have a graphic display with adjustable contrast, an ergonomic keypad and a stainless steel top pan.
  • Selectable filter levels and user can select the measuring unit that the balance will use to display the weight
  • RS232 data interface
  • GLP set-up - stores balance, project and user identification data, as
More Product Information