VWR® Disposable Weighing Boats
Supplier: VWR Collection
611-2249EA 0 INR
611-2249
VWR® Disposable Weighing Boats
Boats Weighing Boats
These flexible PS boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
  • Shallow boat with a wide, flat bottom to resist tipping
  • Temperature range –10 to 70 °C (14 to 158 °F)
  • Can be used with microwave moisture analysers
  • All are FDA/food-grade compliant
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
These sturdy weighing boats are ideal for weighing solid products.