Oxygen cylinder, emptied/unwashed

Supplier: VWR Chemicals
Warning

600823V.EMPTYEA 0 INR
600823V.EMPTY
Oxygen cylinder, emptied/unwashed
Oxygen cylinder, emptied/unwashed
Formula: O₂
Storage Temperature: Ambient
CAS Number: 7782-44-7
EINECS: 231-956-9
UN: 1072
ADR: 2
Merck Index: 12,07098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR