Antistatic explosion proof tubing, b.safe, Bohlender
Supplier: Bohlender
R770-02 R770-08 R770-10 R770-12 R770-14 R770-16
590-0352EA 0 INR
590-0352 590-0353 590-0354 590-0355 590-0356 590-0357
Antistatic explosion proof tubing, b.safe, Bohlender
Tubing Transfer Tubing
Excellent electric conductivity due to a special 'antistatic compound' made of pure PTFE and finest, highly pure carbon dust (less than 2,5%). Colour: black.
  • Extensive chemical resistance due to PTFE parts
  • Resistance of less than 1000 kOhm according to DIN EN 12115 directive
  • Ideal for light-sensitive substances
(1) Bending radius: minimum bending radius in mm at a room temperature of 23 °C
(2) Burst pressure: computed value in bar at a room temperature of 23 °C. It is recommended to restrict the maximum working pressure to 25% of the burst pressure.More Product Information