Waste caps, b.safe
Supplier: Bohlender
M340-01 M340-03 M345-01 M345-10 M345-25 M345-35 M350-01 M350-03 M350-40 M351-01 M351-03 M351-25 M353-01 M353-25 M353-35 M355-01 M355-03 M355-25 M355-35 M355-40 M360-01 M360-03 M360-25 M360-35 M360-40 M363-01 M363-03 M363-30 M365-01 M365-03 M371-01 M371-03 M371-25 M383-01 M390-01 M390-03
590-0110EA 0 INR
590-0110 590-0111 590-0112 590-0114 590-0115 590-0116 590-0117 590-0118 590-0119 590-0120 590-0121 590-0122 590-0123 590-0124 590-0125 590-0126 590-0127 590-0128 590-0129 590-0130 590-0131 590-0132 590-0133 590-0134 590-0135 590-0136 590-0137 590-0138 590-0139 590-0140 590-0141 590-0142 590-0143 590-0144 590-0145 590-0146
Waste caps, b.safe
Closures Safety Caps and Safety Waste Caps
Safe collection of hazardous waste is an important part of the laboratory work routine. b.safe waste caps connect waste lines from your HPLC/UHPLC leak-tight with your waste container. The waste caps are available in various sizes to fit your specific waste container safely.
  • Available in many thread sizes to fit a wide range of waste containers
  • Excellent chemical resistance
  • Accessories like fittings and hose connectors guarantee you waste lines are connected leak tight
  • Special couplings for corrugated flexible tubes
  • Exhaust filters for different sizes of
More Product Information