Hydrochloric acid 20%, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

5853.1000EA 0 INR
5853.1000
Hydrochloric acid 20%, AnalaR NORMAPUR® zA
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Assay (titr.) Min. 20 %
Phosphate Max. 0.00005 %
Sulfate Max. 0.0001 %
Bromide Max. 0.005 %
As Max. 0.000001 %
Ni Max. 0.000002 %
PB Max. 0.000002 %
Al Max. 0.000005 %
Zn Max. 0.000005 %
Fe Max. 0.00002 %
Ca Max. 0.00005 %
NA Max. 0.00005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR