Sulphuric acid 5 N aqueous solution, BAKER ANALYZED® Volumetric, J.T.Baker®

Supplier: Avantor
Danger


M.O.C. Category NI

5696-02EA 11401.19 INR
5696-02 5696-03
Sulphuric acid 5 N aqueous solution, BAKER ANALYZED® Volumetric, J.T.Baker®
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 4.95 - 5.05
Phosphate (PO₄) ≤1 ppm
Heavy Metals (as Pb) ≤0.3 ppm
Chloride (Cl) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR