Sulphuric acid 0.1 N aqueous solution, BAKER ANALYZED® Volumetric, J.T.Baker®

Supplier: Avantor
Danger


M.O.C. Category NI

5641-02EA 6218.38 INR
5641-02 5641-03
Sulphuric acid 0.1 N aqueous solution, BAKER ANALYZED® Volumetric, J.T.Baker®
Sulphuric acid

Standardized at 25°C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to +/- 1 part per 1000, provided on label.

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 0.0995 - 0.1005
Normality 0.0995 - 0.1005
Chloride (Cl) ≤1 ppm
Phosphate (PO₄) ≤1 ppm
Heavy Metals (as Pb) ≤0.3 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR