VWR®, Short Thread Vials and Closures for HPLC, ND9
Supplier: VWR Collection
548-2387EA 637.5 INR
548-2387 548-2389 548-2391 548-2461 548-2462 548-2463 548-2464 548-2465 548-2466 548-2467 548-2468 548-2472 548-2473 548-2474 548-2475 548-2476 548-2477 548-2478 548-2479 548-2545 548-2546 548-2553 548-2580
VWR®, Short Thread Vials and Closures for HPLC, ND9
Vials
USP Type-1 borosillicate glass vials , PP caps with PTFE/Silicone septa, bonded/non bonded, compatible universally on all HPLC autosampleres, prefered type vials for waters and Agilent systems.
  • Special short thread cap for LC-MS and GC-MS applications
  • One component closure - bonded
  • No bleeding
  • Completely inert
Pre-cut closures are available, where only the silicone/PTFE is slitted slightly as per major instrument manufacturer's requirement. Random batch leach free tested in HPLC/MS (report can be submitted on request).
Certifications: Each packMore Product Information