Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
81301 81317 81320 81330 81343 81420 81517 81520 81620 81630 7649-01 81430 81530 85020 86020 82520 81365 81560 CAL81520 CAL81317 CAL81301 CAL81420 CAL81620 CAL85021
549-1189EA 0 INR
549-1189 549-1190 613-1321 549-0369 549-1191 613-1322 613-1303 613-1323 613-1324 549-1267 548-2429 549-1265 549-1266 613-1326 613-1327 548-1971 613-1166 548-2321 548-2352 548-2347 548-2345 548-2350 548-2353 548-2449
Gastight® 1000 series syringes
Syringes Syringes for Chromatography
The 1000 series is a mid volume Gastight® syringe. Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
  • Ideal for dispensing both liquids and gases in volumes from 100 µl up to 100 ml
Removable needle (RN): The designMore Product Information