VWR®, Centrifuge Tubes, ASTM
Supplier: VWR Collection
525-1568EA 0 INR
525-1568 525-1569
VWR®, Centrifuge Tubes, ASTM
Tubes Centrifuge Tubes
Borosilicate glass 3.3, used for the determination of water and sediments in crude oils according to test method ASTM D 96.
  • High chemical and thermal resistance
  • High contrast blue enamel graduations
  • Withstand forces up to 700 ×g
Design, graduations and tolerances meet ASTM (American Society for Testing Materials) specifications.
Certifications:
ASTM E-982
Delivery information: Delivered without cap.