VWR®, Centrifuge Tubes, Metal-Free, Sterile
Supplier: VWR Collection
89049-170 89049-174 89049-172
VWRU89049-170EA 18210 INR
VWRU89049-170 VWRU89049-174 VWRU89049-172 525-1121 525-1123 525-1124 525-1125
VWR®, Centrifuge Tubes, Metal-Free, Sterile
Tubes Centrifuge Tubes
These leakproof centrifuge tubes are ideal for environmental uses, such as water analysis and other applications wherein tubes have the potential to contaminate samples.
  • Easy-to-read black graduations in ±2% increments
  • Contains a large, white frosted writing area
  • RNase-, DNase-, and endotoxin-free
  • Autoclavable (121 °C) and freezable (–80 °C)
  • Can be centrifuged up to 12500×g
  • Sterile tubes are sterilized by irradiation
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
More Product Information