VWR®, Microtube Storage Boxes
Supplier: VWR Collection
525-0925EA 6915.82 INR
525-0925 525-0926 525-0927 525-0928 525-0929 525-0930 525-0931 525-0932 525-0933 525-0934 525-0935 525-0936 525-0937 525-0938 525-0939 525-0940 525-0941
VWR®, Microtube Storage Boxes
Boxes Cryoboxes
PP.
  • Highly transparent, with bold identifying colours and a distinctive wave design
  • Uniform wells with location ID; capacity indicated on the outside of the box
  • Black light-tight version available for light-sensitive samples
  • Frosted labelling patch
  • Alphanumeric identification on the bottom
Compact footprint for microtube and cryogenic vial storage. Store 1,5/2,0 ml microtubes or cryogenic vials. Complete with an easy open, friction-fit lid. Will fit into most standard freezer racking.
Delivery information: Assorted pack contains one of each colourMore Product Information