VWR®, Bottle-Top Vacuum Filtration Systems, High Performance
Supplier: VWR Collection
514-0348EA 0 INR
514-0348 514-0350 514-0349 514-0354 514-0355 514-0358 514-0359 514-0360 514-0362 514-0363 514-0364 514-0366 514-0367 514-0296 514-0297 514-0298 514-0299 514-0300 514-0301 514-0302 514-0303 514-0304 514-0305 514-0306 514-0307
VWR®, Bottle-Top Vacuum Filtration Systems, High Performance
Filters Bottle Top Filters
Designed for the preparation of buffers, tissue culture media, microbiological media and other biological fluids. These vacuum filtration systems feature large diameter high performance PES membranes for guaranteed performance and maximum throughput. The full assembly includes a filtration unit and a receiver bottle with an ergonomic shape, moulded finger grips and a patented bottle cap. The centrifuge tube and funnel system allows the user to filter fluids directly into a 15 or 50 ml tube.
  • Quick-Connect™ pedestal base or cradle ring allows for hands-free
More Product Information