VWR® Testing Sieves, 8" Brass Frame, Stainless Steel Wire Cloth
Supplier: VWR Collection
57334-272 57334-280 57334-290 57334-204 57334-206 57334-208 57334-210 57334-212 57334-214 57334-216 57334-220 57334-222 57334-224 57334-226 57334-228 57334-230 57334-234 57334-232 57334-236 57334-238 57334-240 57334-242 57334-244 57334-606 57334-610 57334-612 57334-614 57334-616 57334-618 57334-620 57334-309 57334-310 57334-314 57334-324 57334-326 57334-328 57334-330 57334-199 57334-200 57334-201 57334-202 57334-274 57334-276 57334-278 57334-282 57334-284 57334-286 57334-292 57334-294 57334-296 57334-298 57334-300 57334-302 57334-334 57334-336 57334-339 57334-340 57334-342 57334-344 57334-346 57334-348 57334-350 57334-352 57334-354 57334-356 57334-358 57334-360 57334-246 57334-248 57334-250 57334-252 57334-254 57334-256 57334-258 57334-260 57334-262 57334-264 57334-266 57334-268 57334-270 57334-304 57334-306 57334-308 57334-366 57334-364 57334-368 57334-370 57334-372 57334-374 57334-376 57334-378 57334-380 57334-382 57334-384 57334-622 57334-624 57334-626 57334-628 57334-630 57334-632 57334-634 57334-636
510-1581EA 0 INR
510-1581 510-1582 510-1583 510-1384 510-1385 510-1386 510-1387 510-1388 510-1389 510-1390 510-1391 510-1392 510-1393 510-1394 510-1395 510-1396 510-1398 510-1397 510-1399 510-1400 510-1402 510-1403 510-1404 510-1564 510-1565 510-1566 510-1567 510-1568 510-1569 510-1570 510-1434 510-1435 510-1436 510-1437 510-1438 510-1439 510-1440 510-1380 510-1381 510-1382 510-1383 510-1418 510-1419 510-1420 510-1421 510-1422 510-1423 510-1425 510-1426 510-1427 510-1428 510-1429 510-1430 510-1441 510-1442 510-1443 510-1444 510-1445 510-1446 510-1447 510-1448 510-1449 510-1450 510-1451 510-1452 510-1453 510-1454 510-1405 510-1406 510-1407 510-1408 510-1409 510-1410 510-1411 510-1412 510-1413 510-1414 510-1415 510-1416 510-1417 510-1431 510-1432 510-1433 510-1457 510-1456 510-1458 510-1459 510-1460 510-1461 510-1462 510-1463 510-1464 510-1465 510-1466 510-1571 510-1572 510-1573 510-1574 510-1575 510-1576 510-1577 510-1578
VWR® Testing Sieves, 8" Brass Frame, Stainless Steel Wire Cloth
Sieves Test Sieves
Brass frame and stainless steel wire cloth offers higher durability than all brass construction. Sieves feature sturdy construction with fillet solder bead between the frame and wire cloth to prevent particles from being trapped. Diameter: 20.3cm (8"). Choose full height with 5.08cm (2") depth to wire, or half height with 2,54 cm (1") depth to wire.
Inspected with an optical measurement system traceable to the National Institute of Standards and Technology. Each sieve bears a traceable serial number and comes with certificate of compliance to applicable ASTM E-11 and ISO specifications.